Bis(tri-o-tolylphosphine)palladium(0) (14.9% Pd)
Supplier: Thermo Scientific Chemicals
|
Warning
|
045814-03
045814-MD
AA45814-03EA
324.94
USD
AA45814-03
AA45814-MD
Bis(tri-o-tolylphosphine)palladium(0) (14.9% Pd)
Bis(tri-o-tolylphosphine)palladium(0)
CAS No.: 69861-71-8
Molecular Formula: C42H42P2Pd
Formula Weight: 715.16
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Off-white to yellow
Melting Point: 220-225°
MDL No.: MFCD12911908
Formula:
C₄₂H₄₂P₂Pd Melting Pt: 220…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD12911908 CAS Number: 69861-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...