Cobalt(II,III) oxide ≥99.7% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tricobalt tetraoxide, Cobaltic tetraoxide
045806-0B
045806-18
045806-30
AA45806-0BEA
917
USD
AA45806-0B
AA45806-18
AA45806-30
Cobalt(II,III) oxide ≥99.7% (metals basis)
Cobalt(II,III) oxide
Purity: 99.7%
CAS No.: 1308-06-1
Molecular Formula: Co3O4
Formula Weight: 240.80
Physical Form: 2-6 Micron Powder
Density: 6.11
Appearance: Black
Melting Point: 900° dec.
Merck Reference: 14,2429
UN No.: 3077
MDL No.: MFCD00010939
Formula:
Co₃O₄ MW: 240.8 g/mol Melting Pt: 895 °C Density: 6.005 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010939 CAS Number: 1308-06-1 EINECS: 215-157-2 UN: 3077 ADR: 9,III Merck Index: 13,02454 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...