Silica gel, Pore size: 260 Å ≥99.99%, spherical 3 µm APS for HPLC

Supplier: Thermo Scientific Chemicals
Warning

045804-03 045804-06
AA45804-03EA 76.6 USD
AA45804-03 AA45804-06 AA45804-14
Silica gel, Pore size: 260 Å ≥99.99%, spherical 3 µm APS for HPLC
Silica gel

Purity: 99.99+% CAS No.: 63231-67-4 Physical Form: S.A. 130m2/g, P.V. 0.90cc/g MDL No.: MFCD00011232

Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR