Silica gel, Pore size: 120 Å ≥99.99%, spherical 3 µm APS for HPLC

Supplier: Thermo Scientific Chemicals
Warning

045803-03 045803-06
AA45803-03EA 54.4 USD
AA45803-03 AA45803-06 AA45803-14
Silica gel, Pore size: 120 Å ≥99.99%, spherical 3 µm APS for HPLC
Silica gel

Purity: 99.99+% CAS No.: 63231-67-4 Physical Form: S.A. 340m2/g, P.V. 1.00cc/g MDL No.: MFCD00011232

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR