Silica gel, Pore size: 110 Å ≥99.99%, spherical 1.6 µm APS for HPLC, UHPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
045795-03
AA45795-03EA
113
USD
AA45795-03
Silica gel, Pore size: 110 Å ≥99.99%, spherical 1.6 µm APS for HPLC, UHPLC
Silica gel
Purity: 99.99+%
CAS No.: 63231-67-4
Physical Form: S.A. 340m2/g, P.V. 0.95cc/g
MDL No.: MFCD00011232
Formula:
SiO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...