Povidone iodine
Supplier: Thermo Scientific Chemicals
045782-18
045782-30
045782-A1
AA45782-18EA
67
USD
AA45782-18
AA45782-30
AA45782-A1
Povidone iodine
Povidone iodine
CAS No.: 25655-41-8
Molecular Formula: (C6H9NO)n.xI
Physical Form: Powder
Appearance: Yellow-brown
Melting Point: 300° dec.
MDL No.: MFCD00084483
Formula:
C₆H₉I₂NO Melting Pt: 300 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00084483 CAS Number: 25655-41-8 Merck Index: 14,7698 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...