Nitinol, foil, shape memory, flat annealed, pickled surface, Thickness 0.05 mm (0.002 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
045663-FF
045663-FI
045663-GE
AA45663-FFEA
96.24
USD
AA45663-FF
AA45663-FI
AA45663-GE
Nitinol, foil, shape memory, flat annealed, pickled surface, Thickness 0.05 mm (0.002 in)
Nitinol
Physical Form: ≈0.25g/25x25mm; Af approx. 45°C
Odor: Odorless
MDL No.: MFCD02091734
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...