Silver conductive ink

Supplier: Thermo Scientific Chemicals
Warning

045661-06 045661-14 045661-22
AA45661-06EA 44.35 USD
AA45661-06 AA45661-14 AA45661-22
Silver conductive ink
Silver conductive ink

Physical Form: Liquid S-020 (formerly Mattheylec P2006) Density: 2.5 Appearance: Silver UN No.: 3082 MDL No.: MFCD01868501

Density: 2.5
Storage Temperature: Ambient
MDL Number: MFCD01868501
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR