Polyethyleneimine, M.W. 1,200 ≥99%, branched

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)

045594-22 045594-36 045594-14
AA45594-22EA 189.91 USD
AA45594-22 AA45594-36 AA45594-14
Polyethyleneimine, M.W. 1,200 ≥99%, branched
Polyethyleneimine

Purity: 99% CAS No.: 9002-98-6 Molecular Formula: (-NHCH2CH2-)x(-N(CH2CH2NH2)CH2CH2-)y Physical Form: Liquid Appearance: Pale yellow Odor: Amine-like MDL No.: MFCD00803910

Formula: (CH2CH2NH)n
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR