2,3,4,6-Tetra-O-benzyl-D-galactopyranose ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(3R,4S,5S,6R)-3,4,5-Tris(benzyloxy)-6-((benzyloxy)methyl)tetrahydro-2H-pyran-2-ol, 2,3,4,6-Tetra-O-benzyl-D-galactose
045531-03
045531-06
AA45531-03EA
105
USD
AA45531-03
AA45531-06
2,3,4,6-Tetra-O-benzyl-D-galactopyranose ≥98%
2,3,4,6-Tetra-O-benzyl-D-galactopyranose
Purity: 98%
CAS No.: 53081-25-7
Molecular Formula: C34H36O6
Formula Weight: 540.65
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline
Appearance: White
MDL No.: MFCD00132927
Formula:
C₃₄H₃₆O₆ MW: 540.65 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00132927 CAS Number: 6386-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...