Nitinol, foil, shape memory, flat annealed, pickled surface, Thickness 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
045514-FF
045514-FI
045514-GH
AA45514-FFEA
81.7
USD
AA45514-FF
AA45514-FI
AA45514-GH
Nitinol, foil, shape memory, flat annealed, pickled surface, Thickness 0.127 mm (0.005 in)
Nitinol
Physical Form: ≈0.51g/25x25mm; Af approx. 45°C
Odor: Odorless
MDL No.: MFCD02091734
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...