Bismuth(III) 2-ethylhexanoate ∼92% in 2-ethylhexanoic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bismuth 2-ethylhexanoate
045500-14
045500-06
045500-22
AA45500-14EA
94.3
USD
AA45500-14
AA45500-06
AA45500-22
Bismuth(III) 2-ethylhexanoate ∼92% in 2-ethylhexanoic acid
Bismuth (III) 2-ethylhexanoate
Purity: 92%
CAS No.: 67874-71-9
Molecular Formula: C24H45BiO6
Formula Weight: 638.61
Physical Form: Liquid
Formula:
C₂₄H₄₅BiO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00043803 CAS Number: 67874-71-9 EINECS: 267-499-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...