Silver behenate
Supplier: Thermo Scientific Chemicals
Synonyms:
Docosanoic Acid Silver Salt, Behenic Acid Silver Salt, Silver Docosanoate
045494-06
045494-14
AA45494-06EA
106.72
USD
AA45494-06
AA45494-14
Silver behenate
Silver behenate
CAS No.: 2489-05-6
Molecular Formula: C22H43O2Ag
Formula Weight: 447.44
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Melting Point: dec.
MDL No.: MFCD00059001
Formula:
C₂₂H₄₄O₂ MW: 447.45 g/mol Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00059001 CAS Number: 2489-05-6 EINECS: 219-641-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...