Methyl cellulose, 15 cPs
Supplier: Thermo Scientific Chemicals
Synonyms:
Cellulose methyl ether
045490-36
045490-A4
045490-22
AA45490-36EA
120
USD
AA45490-36
AA45490-A4
AA45490-22
Methyl cellulose, 15 cPs
Methyl cellulose
CAS No.: 9004-67-5
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD00081763
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 Merck Index: 14,6040 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...