Tris(1-methoxy-2-methyl-2-propoxy)bismuth ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
045488-14 045488-06
AA45488-14EA 1374 USD
AA45488-14 AA45488-03 AA45488-06
Tris(1-methoxy-2-methyl-2-propoxy)bismuth ≥99.99% (metals basis)
Tris(1-methoxy-2-methyl-2-propoxy)bismuth
Purity: 99.99% Molecular Formula: C15H33O6Bi Formula Weight: 518.40 Physical Form: Crystalline, Packaged under argon Appearance: Yellow Melting Point: 70° MDL No.: MFCD11113030