Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Diameter 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

045086-G1 045086-G5
AA45086-G6EA 896.49 USD
AA45086-G6 AA45086-G1 AA45086-G5
Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, wire, Diameter 0.1 mm (0.004 in)
Tungsten (gold coated)

Purity: 99.95% Physical Form: ≈0.15g/m (W only), 3-5 wt% Au plating MDL No.: MFCD00011461

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR