Tetraamminepalladium(II) hydrogen carbonate

Supplier: Thermo Scientific Chemicals
Danger

045082-03 045082-06
AA45082-03EA 150 USD
AA45082-03 AA45082-06
Tetraamminepalladium(II) hydrogen carbonate
Tetraamminepalladium(II) hydrogen carbonate

CAS No.: 134620-00-1 Molecular Formula: C2H14N4O6Pd Formula Weight: 296.58 Physical Form: Crystalline Density: 2.04 Appearance: Colorless to light yellow UN No.: 3077 MDL No.: MFCD09953449

Formula: C₂H₁₄N₄O₆Pd
MW: 296.58 g/mol
Density: 2.04
Storage Temperature: Ambient
MDL Number: MFCD09953449
CAS Number: 134620-00-1
EINECS: 425-270-0
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR