Poly(styrene sulfonic acid) sodium salt, M.W. 500,000
Supplier: Thermo Scientific Chemicals
Synonyms:
Poly(sodium-p-styrenesulphonate)
045050-14
045050-22
AA45050-14EA
62.33
USD
AA45050-14
AA45050-22
Poly(styrene sulfonic acid) sodium salt, M.W. 500,000
Poly(styrene sulfonic acid) sodium salt
CAS No.: 25704-18-1
Molecular Formula: [-CH2CH(C6H4)SO3Na)-]n
Physical Form: Powder
Appearance: White to pale brown
MDL No.: MFCD00084449
Formula:
[-CH₂CH(C₆H₄)SO₃Na)-]n Storage Temperature: Ambient |
MDL Number:
MFCD00084449 CAS Number: 25704-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...