[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride (17,6% Pd)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,4-Butylenebis(diphenylphosphine)palladium dichloride, Dichloro[1,4-bis(diphenylphosphino)butane]palladium(II)
044971-06
044971-03
AA44971-06EA
868
USD
AA44971-06
AA44971-03
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride (17,6% Pd)
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride
CAS No.: 29964-62-3
Molecular Formula: C28H28Cl2P2Pd
Formula Weight: 603.80
Physical Form: Lump
MDL No.: MFCD02093437
Formula:
C₂₈H₂₈Cl₂P₂Pd MW: 603.80 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02093437 CAS Number: 29964-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...