Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Supplier: Thermo Scientific Chemicals
|
Warning
|
044955-03
044955-06
044955-14
AA44955-03EA
114
USD
AA44955-03
AA44955-06
AA44955-14
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
CAS No.: 16971-33-8
Molecular Formula: C55H46ClOP3Ru
Formula Weight: 952.41
Physical Form: Crystalline
Sensitivity: Moisture and Light Sensitive
Appearance: Gray
Melting Point: 204-206° dec.
MDL No.: MFCD00049804
Formula:
C₅₅H₄₅ClOP₃Ru MW: 951.405 g/mol Melting Pt: 204…206 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00049804 CAS Number: 16971-33-8 EINECS: 241-051-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...