Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
044953-FF
044953-FI
AA44953-FFEA
136
USD
AA44953-FF
AA44953-FI
Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness 0.25 mm (0.01 in)
Nitinol
Physical Form: ≈1.02g/25x25mm; Af approx. 10°C
Odor: Odorless
MDL No.: MFCD02091734
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...