Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
044952-FF
044952-FI
AA44952-FFEA
87.54
USD
AA44952-FF
AA44952-FI
Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness 0.127 mm (0.005 in)
Nitinol
Physical Form: ≈0.51g/25x25mm; Af approx. 10°C
Odor: Odorless
MDL No.: MFCD02091734
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...