Tetramethylammonium hydroxide 1.0 M in aqueous solution ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
044938-AK
044938-K2
AA44938-AKEA
69.16
USD
AA44938-AK
AA44938-K2
Tetramethylammonium hydroxide 1.0 M in aqueous solution ACS
Tetramethylammonium hydroxide
CAS No.: 75-59-2
Molecular Formula: C4H13NO
Formula Weight: 91.15
Physical Form: Liquid
Sensitivity: Air Sensitive
Appearance: Colorless to pale yellow
Beilstein Reference: 3558708
Merck Reference: 14,9224
UN No.: 1835
MDL No.: MFCD00008280
Formula:
C₄H₁₃NO MW: 91.15 g/mol Boiling Pt: 110 °C (1013 hPa) Density: 1.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008280 CAS Number: 75-59-2 EINECS: 200-882-9 UN: 1835 ADR: 8,III Merck Index: 14,9224 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...