Indium tin oxide ≥99.5%, NanoTek®
Supplier: Thermo Scientific Chemicals
Synonyms:
ITO
NanoTek® indium tin oxide can be used to impart conductivity to transparent coatings such as those on the surface of computer display tubes or on the films used to produce conductive display screens such as for interactive touch screens
Applications: Indium tin oxide is one of the most widely used transparent conducting oxides due to its important properties like electrical conductivity and optical transparency, as well as the easy formation of thin film on deposition. It is also used in touch panels, electrochromic, electroluminescent and LCD displays, plasma displays, field emission displays, heat reflective coatings, energy efficient windows, gas sensors and photovoltaics.
Applications: NanoTek™ indium tin oxide can be used to impart conductivity to transparent coatings such as those on the surface of computer display tubes or on the films used to produce conductive display screens such as for interactive touch screens
Formula:
In₂O₃·(SnO₂)x Density: 7,16 Storage Temperature: Ambient |
MDL Number:
MFCD00171662 CAS Number: 50926-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...