Poly(3-hexylthiophene-2,5-diyl) regioregular low metals
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
P3HT, Poly(3-hexylthiophene), Poly[3-hexylthiophene-2,5-diyl]
044745-77
044745-02
AA44745-77EA
116
USD
AA44745-77
AA44745-02
Poly(3-hexylthiophene-2,5-diyl) regioregular low metals
Poly(3-hexylthiophene-2,5-diyl)
Soluble in Chloroform and tetrahydrofuran
CAS No.: 104934-50-1
Physical Form: Powder
Solubility: Soluble in Chloroform and tetrahydrofuran
MDL No.: MFCD00217686
MW:
196.352 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00217686 CAS Number: 104934-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...