Chromium etchant

Supplier: Thermo Scientific Chemicals
Danger

044582-R0 044582-LV
AA44582-R0EA 190.59 USD
AA44582-R0 AA44582-LV
Chromium etchant
Chromium etchant

Applications: High purity ceric ammonium nitrate system for precise, clean etching of chromium and chromium oxide films


Physical Form: Liquid Density: 1.16 Appearance: Orange Odor: Acrid UN No.: 3093 MDL No.: MFCD07370678


Applications: High purity ceric ammonium nitrate system for precise, clean etching of chromium and chromium oxide films

Density: 1.16
Storage Temperature: Ambient
MDL Number: MFCD07370678
UN: 3093
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR