Calcium-α-D-isosaccharinate ≥98%
Supplier: Thermo Scientific Chemicals
044371-14
044371-03
044371-06
AAA44371-01EA
34.72
USD
AAA44371-01
AA44371-14
AA44371-03
AA44371-06
Calcium-α-D-isosaccharinate ≥98%
Calcium-alpha-D-isosaccharinate
Purity: 98%
Molecular Formula: C12H22CaO12
Formula Weight: 398.38
Physical Form: Crystalline
Appearance: Pale brown
MDL No.: MFCD04039767
Specification Test Results
Form | Crystalline |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...