PTFE 6-10 micron
Supplier: Thermo Scientific Chemicals
044184-A1
044184-18
044184-30
AA44184-A1EA
910
USD
AA44184-A1
AA44184-18
AA44184-30
PTFE 6-10 micron
PTFE
CAS No.: 9002-84-0
Molecular Formula: -(CF2CF2-)n
Physical Form: Powder
MDL No.: MFCD00084455
Formula:
(CF₂CF₂)n Storage Temperature: Ambient |
MDL Number:
MFCD00084455 CAS Number: 9002-84-0 Merck Index: 14,7583 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...