Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III)

Supplier: Thermo Scientific Chemicals

044175-02 044175-77
AA44175-02EA 386 USD
AA44175-02 AA44175-77
Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III)
Tris(1,3-diphenyl-1,3-propanedionato)(1,10-phenanthroline)europium(III)

Common Applications: as an organic red light emitter
œex 369nm, œem 612nm
Soluble in dichloromethane, THF


CAS No.: 17904-83-5 Molecular Formula: C57H41EuN2O6 Formula Weight: 1001.93 Physical Form: Powder Melting Point: 172-173° Solubility: Soluble in dichloromethane, THF MDL No.: MFCD01321202


Applications: As an organic red light emitter

Formula: C₅₇H₄₁EuN₂O₆
MW: 1001.93 g/mol
Melting Pt: 172…173 °C
Storage Temperature: Ambient
MDL Number: MFCD01321202
CAS Number: 17904-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR