Silver plating solution ∼2.1 g/l metal content, electroless
Supplier: Thermo Scientific Chemicals
|
Danger
|
044068-AK
044068-K2
AA44068-AKEA
54.4
USD
AA44068-AK
AA44068-K2
Silver plating solution ∼2.1 g/l metal content, electroless
Silver plating solution
Applications: For plating on copper, brass or copper alloys
Physical Form: Liquid
Density: 1.04
Appearance: Colorless
Melting Point: -5°
Boiling Point: 103-105°
UN No.: 3287
MDL No.: MFCD00003397
Applications: For plating on copper, brass or copper alloys
Boiling Pt:
103…105 °C Melting Pt: –5 °C Density: 1.04 Storage Temperature: Ambient |
MDL Number:
MFCD00003397 UN: 3287 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...