Silver plating solution ∼28.7 g/l (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
044067-K2
044067-AK
AA44067-K2EA
154
USD
AA44067-K2
AA44067-AK
Silver plating solution ∼28.7 g/l (metals basis)
Silver plating solution
UN No.: 1935
MDL No.: MFCD00003397
Storage Temperature:
Ambient |
MDL Number:
MFCD00003397 UN: 1935 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...