Magnesium tetraborate ≥99%
Supplier: Thermo Scientific Chemicals
043892-22
043892-14
AA43892-22EA
427
USD
AA43892-22
AA43892-14
Magnesium tetraborate ≥99%
Magnesium tetraborate
Purity: 99%
CAS No.: 12007-62-4
Molecular Formula: MgB4O7
Formula Weight: 179.54
Physical Form: Powder
Odor: Odorless
MDL No.: MFCD02688001
Formula:
MgB₄O₇ Storage Temperature: Ambient |
MDL Number:
MFCD02688001 CAS Number: 12007-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...