Triiron phosphide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Iron phosphide
043486-09
043486-18
AA43486-09EA
163
USD
AA43486-09
AA43486-18
Triiron phosphide ≥99.5% (metals basis)
Triiron phosphide
Purity: 99.5%
CAS No.: 12023-53-9
Molecular Formula: Fe3P
Formula Weight: 198.51
Physical Form: -40 Mesh Powder
Formula:
Fe₃P MW: 198.51 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00799762 CAS Number: 12023-53-9 EINECS: 234-682-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...