Mesoporphyrin IX dihydrochloride ≥97%

Supplier: Thermo Scientific Chemicals

043320-MA 043320-MF 043320-MD
AA43320-MAEA 37.6 USD
AA43320-MA AA43320-MF AA43320-MD
Mesoporphyrin IX dihydrochloride ≥97%
Mesoporphyrin IX dihydrochloride

Purity: 97% CAS No.: 68938-72-7 Molecular Formula: C24H38N4O4.2HCl Formula Weight: 639.62 Physical Form: Crystalline MDL No.: MFCD00136049

Formula: C₂₄H₃₈N₄O₄.2HCl
Storage Temperature: Ambient
MDL Number: MFCD00136049
CAS Number: 68938-72-7
EINECS: 273-141-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR