Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, annealed, Diameter 0.38 mm (0.015 in) Type 304
Supplier: Thermo Scientific Chemicals
043301-BY
043301-CC
AA43301-BYEA
49.6
USD
AA43301-BY
AA43301-BU
AA43301-CC
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, annealed, Diameter 0.38 mm (0.015 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Temper: soft
Odor: Odorless
MDL No.: MFCD00148499
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...