Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, annealed, Diameter 0.25 mm (0.01 in) Type 304
Supplier: Thermo Scientific Chemicals
Danger
043300-BU 043300-CC 043300-BY
AA43300-BUEA 28 USD
AA43300-BU AA43300-CC AA43300-BY
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, annealed, Diameter 0.25 mm (0.01 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% Physical Form: Temper: soft Odor: Odorless MDL No.: MFCD00148499