Benzyl acrylate 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 150 ppm

043204-18 043204-30 043204-09
AA43204-18EA 154 USD
AA43204-18 AA43204-30 AAA43204-09
Benzyl acrylate 98% stabilized
Benzyl acrylate

Stab. with ca 150ppm 4-methoxyphenol Preparation of high refractive index polyacrylates Soluble in organic solvents


Applications: Preparation of high refractive index polyacrylates


Applications: Preparation of high refractive index polyacrylates

Formula: C₁₀H₁₀O₂
Boiling Pt: 111 °C (8 mmHg)
Density: 1.06
Storage Temperature: Ambient
MDL Number: MFCD00048147
CAS Number: 2495-35-4
EINECS: 219-673-9
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR