Poly(vinylbenzyl chloride)
Supplier: Thermo Scientific Chemicals
|
Warning
|
043067-04
043067-18
043067-09
AAA43067-04EA
81.2
USD
AAA43067-04
AA43067-18
AA43067-09
Poly(vinylbenzyl chloride)
Poly(vinylbenzyl chloride)
M.W. range 20,000-50,000
CAS No.: 9080-67-5
Molecular Formula: -[CH2CH(C6H4CH2Cl-]n
Physical Form: Solid
MDL No.: MFCD00135445
Formula:
-[CH₂CH(C₆H₄CH₂Cl-]n Storage Temperature: Ambient |
MDL Number:
MFCD00135445 CAS Number: 9080-67-5 |
Specification Test Results
Form | Solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...