Lithium tri-sec-butylborohydride 1.0 M in THF

Supplier: Thermo Scientific Chemicals
Danger


Packaged under Argon in resealable ChemSeal™ bottles.

043034-AE 043034-AP
AA43034-AEEA 114.75 USD
AA43034-AE AA43034-AP
Lithium tri-sec-butylborohydride 1.0 M in THF
Lithium tri-sec-butylborohydride

Packaged under Argon in resealable ChemSeal™ bottles Uses: reducing agent of olefinic groups and ketones


Applications: As a reducing agent of olefinic groups and ketones


Applications: As a reducing agent of olefinic groups and ketones

Formula: C₁₂H₂₇BLi
Density: 0.870
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011708
CAS Number: 38721-52-7
UN: 3394
ADR: 4.2,I
Merck Index: 14,8427

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR