Tetrakis(dimethylamido)titanium(IV) ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Titanium(IV) dimethylamide, TDMAT, Tetrakis(dimethylamino)titanium(IV)
042932-06
042932-14
AA42932-06EA
201
USD
AA42932-06
AA42932-14
Tetrakis(dimethylamido)titanium(IV) ≥99.9% (metals basis)
Tetrakis(dimethylamino)titanium (TDMAT)
Purity: 99.9%
CAS No.: 3275-24-9
Molecular Formula: C8H24N4Ti
Formula Weight: 224.19
Flash Point: -30°(-22°F)
Physical Form: Liquid
Density: 0.96
Sensitivity: Moisture Sensitive
Appearance: Yellow-brown
Boiling Point: 50°/0.5mm
UN No.: 3129
MDL No.: MFCD00014861
Formula:
C₈H₂₄N₄Ti MW: 224.17 g/mol Boiling Pt: 50 °C (0.5 mmHg) Density: 0.96 Flash Pt: –30 °C (–22 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014861 CAS Number: 3275-24-9 EINECS: 221-904-3 UN: 3129 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...