Tetraammineplatinum(II) hydroxide solution (8 - 11% Pt) (w/w)

Supplier: Thermo Scientific Chemicals
Warning

042918-NH 042918-NB
AA42918-NHEA 315 USD
AA42918-NH AA42918-NB
Tetraammineplatinum(II) hydroxide solution (8 - 11% Pt) (w/w)
Tetraammineplatinum(II) hydroxide

CAS No.: 38201-97-7 Molecular Formula: H14N4O2Pt Formula Weight: 297.23 Physical Form: Liquid Appearance: Colorless Odor: Odorless MDL No.: MFCD00050044

Formula: H₁₄N₄O₂Pt
MW: 297.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050044
CAS Number: 38201-97-7
EINECS: 253-823-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR