Scandium(III) sulfate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

042877-03 042877-06
AA42877-03EA 161 USD
AA42877-03 AA42877-06
Scandium(III) sulfate hydrate ≥99.9% (REO, rare earth oxide basis)
Scandium(III) sulfate hydrate

Freely soluble in water


Purity: 99.9% Molecular Formula: Sc2(SO4)3.XH2O Formula Weight: 378.10 (anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Solubility: Freely soluble in water MDL No.: MFCD00149854

Formula: Sc₂(SO₄)₃.XH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149855
CAS Number: 52788-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR