Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 50% water

Supplier: Thermo Scientific Chemicals

042578-04 042578-09 042578-18
AA42578-04EA 140.94 USD
AA42578-04 AA42578-09 AA42578-18
Palladium hydroxide (20% Pd) on carbon (Pearlman's catalyst) wetted with ca. 50% water
Palladium hydroxide

Uses: Hydrogenolysis of benzyl-nitrogen bonds
Sold on a dry weight basis. Unit weight excludes water weight


CAS No.: 12135-22-7 Odor: Odorless MDL No.: MFCD00064599


Applications: Hydrogenolysis of benzyl-nitrogen bonds

Formula: H₂O₂Pd
MW: 140.435 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064599
CAS Number: 12135-22-7
EINECS: 235-219-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR