Apatite naturally occurring mineral, grains ∼ 0.06-0.19 in

Supplier: Thermo Scientific Chemicals
Warning

042535-03 042535-06 042535-14
AA42535-03EA 35.5 USD
AA42535-03 AA42535-06 AA42535-14
Apatite naturally occurring mineral, grains ∼ 0.06-0.19 in
Apatite

Apatite is one of a few minerals produced and used by biological micro-environmental systems. The primary use of apatite is in the manufacture of fertilizer, it is a source of phosphorus. It is occasionally used as a gemstone. Green and blue varieties in finely divided form, are pigments with excellent covering power.


Sp. Gr. 3.1-3.2. Store away from strong oxidizing agents. Store in cool, dry conditions.


Applications: Grain mounts, XRD and microprobe standards, identification of unknown minerals

Formula: Ca₅FO₁₂P₃
Storage Temperature: Ambient
MDL Number: MFCD00285511
CAS Number: 1306-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR