N,N-Dimethylacrylamide ≥99.5% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: 100 ppm

042405-18 042405-30 042405-A1
AA42405-18EA 44.84 USD
AA42405-18 AA42405-30 AA42405-A1
N,N-Dimethylacrylamide ≥99.5% stabilized
N,N-Dimethylacrylamide

Stab. with 100ppm 4-methoxyphenol


Purity: 99.5% CAS No.: 2680-03-7 Molecular Formula: C5H9NO Formula Weight: 99.13 Flash Point: 71°(161°F) Physical Form: Liquid Density: 0.962 Refractive Index: 1.4730 Sensitivity: Light Sensitive Appearance: Colorless Boiling Point: 82-84°/21mm Beilstein Reference: 1742219 UN No.: 2810 MDL No.: MFCD00008626

Formula: C₅H₉NO
Boiling Pt: 82…84 °C (21 mmHg)
Density: 0.962
Flash Pt: 71 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008626
CAS Number: 2680-03-7
EINECS: 220-237-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR