Poly(2-ethyl-2-oxazoline), M.W. ~200,000

Supplier: Thermo Scientific Chemicals

042397-A4 042397-22 042397-36
AA42397-A4EA 401 USD
AA42397-A4 AA42397-22 AA42397-36
Poly(2-ethyl-2-oxazoline), M.W. ~200,000
Poly(2-ethyl-2-oxazoline), M.W. ~200,000

Nominal M.W. 200,000


CAS No.: 25805-17-8 Molecular Formula: [-N(COC2H5)CH2CH2-]n Physical Form: Solid MDL No.: MFCD00134377

Formula: (C₅H₉NO)x
MW: 200000 g/mol
Storage Temperature: Ambient
CAS Number: 25805-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR