Aluminum silicon slug (99:1 wt%) ≥99.99% (metals basis), Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
042322-18
042322-09
042322-30
AA42322-18EA
115
USD
AA42322-18
AA42322-09
AA42322-30
Aluminum silicon slug (99:1 wt%) ≥99.99% (metals basis), Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Aluminum silicon slug
Purity: 99.99%
CAS No.: 11145-30-5
Molecular Formula: Al:Si; 99:1 wt%
Odor: Odorless
MDL No.: MFCD00143595
Storage Temperature:
Ambient |
MDL Number:
MFCD00143595 CAS Number: 11145-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...