Indium oxide, tin doped (∼20% Oxide) (w/w) polymeric precursor
Supplier: Thermo Scientific Chemicals
|
Danger
|
042229-09
042229-14
AA42229-04EA
983.67
USD
AA42229-04
AA42229-09
AA42229-14
Indium oxide, tin doped (∼20% Oxide) (w/w) polymeric precursor
Indium oxide, tin doped
Applications: Preparation of transparent electrical conducting electrodes
CAS No.: 1312-43-2
Physical Form: Viscous liquid
Solubility: Soluble in hexane and toluene. Insoluble in alcohols
UN No.: 1993
MDL No.: MFCD00011060
Applications: Preparation of transparent electrical conducting electrodes
Flash Pt:
13 °C (55 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011060 CAS Number: 1312-43-2 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...