Nickel plating solution, semi-bright finish
Supplier: Thermo Scientific Chemicals
|
Danger
|
042026-K2
042026-AK
AA42026-K2EA
72.7
USD
AA42026-K2
AA42026-AK
Nickel plating solution, semi-bright finish
Nickel plating solution
Physical Form: Liquid
Appearance: Green
UN No.: 3287
MDL No.: MFCD00011137
Storage Temperature:
Ambient |
MDL Number:
MFCD00011137 UN: 3287 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...