Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 150 mesh woven from 0.066mm (0.0026in) Type 304
Supplier: Thermo Scientific Chemicals
042010-RF
042010-RU
AA42010-RFEA
154.43
USD
AA42010-RF
AA42010-RU
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire 150 mesh woven from 0.066mm (0.0026in) Type 304
Stainless steel
Open area: 37.2%; Width of opening: 0.0041in
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Wire Cloth
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...